Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Synuclein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589862 Supplier Invitrogen™ Supplier No. PA589862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSVTSVAEKT KEQANAVSEA VVSSVNTVAT KTVEEAENIA VTSGVVRKED LRPSAPQQEG EASKEKEEVA EEAQSGGD; Positive Samples: SKOV3, HT-29; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, perinuclear region, spindle.

This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Synuclein
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNCG
Gene Accession No. O76070, Q9Z0F7
Gene Alias BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1)
Gene Symbols SNCG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-127 of human gamma-Synuclein (NP_0030782).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20618, 6623
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.