Learn More
Gamma-Taxilin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304043AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the taxilin family. The encoded protein binds to the C-terminal coiled-coil region of syntaxin family members 1A, 3A and 4A, and may play a role in intracellular vesicle trafficking. This gene is up-regulated by lipopolysaccharide and the gene product may be involved in cell cycle regulation. The related mouse protein was also shown to inhibit activating transcription factor 4-mediated transcription and thus regulate bone mass accrual. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| Gamma-Taxilin | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8BHN1, Q9NUQ3 | |
| TXLNG | |
| Between 478 and 528 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TXLNG | |
| taxilin gamma, CXorf15, ELRG, FIAT, LSR5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353170, 55787 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.