Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gamma-taxilin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980404
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0404 Supplier Bioss Supplier No. BS0631R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.

Specifications

Antigen Gamma-taxilin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TXLNG
Gene Accession No. Q9NUQ3
Gene Alias 4932441K18Rik; BOS_26341; CXHXORF15; CXorf15; ELRG; Environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; Factor inhibiting ATF4-mediated transcription; FIAT; Gamma-taxilin; lipopolysaccaride-specific response 5-like protein-like; lipopolysaccharide specific response-5 protein; Lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; LOC100352103; Lrg; Lrp; LSR5; taxilin gamma; Txlng; TXLNGX
Gene Symbols TXLNG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Gamma-taxilin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55787
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.