Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Tubulin Antibody (4D11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10074421
Description
gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
gamma Tubulin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 | |
gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
gamma Tubulin (4D11) | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
4D11 | |
Unconjugated | |
P23258 | |
TUBG1 | |
Recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin. | |
100 μg | |
Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers | |
7283 | |
Human, Mouse, Rat, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction