Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Tubulin Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299067MFV610
Description
gamma Tubulin Polyclonal antibody specifically detects gamma Tubulin in Human samples. It is validated for Western BlotSpecifications
gamma Tubulin | |
Polyclonal | |
Western Blot | |
gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human gamma Tubulin. | |
0.1 mL | |
Cell Biology, Cellular Markers, Centrosome Markers | |
7283 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction