Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Tubulin Antibody (TU-30), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500574MFV500
Description
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human, Mouse, Rat, Porcine, Bovine, Chicken, Plant, Protozoa samples. It is validated for Flow CytometrySpecifications
gamma Tubulin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Chicken, Plant | |
Purified |
Flow Cytometry | |
TU-30 | |
Flow Cytometry | |
gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
C-terminal peptide of gamma-tubulin counjugated to KLH. | |
0.1 mL | |
Cell Biology, Cellular Markers, Centrosome Markers | |
7283 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction