Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma tubulin Centrosome Marker Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2722 Supplier Bioss Supplier No. BS1322R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.

Specifications

Antigen gamma tubulin Centrosome Marker
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tubg2
Gene Accession No. P23258, Q9NRH3
Gene Alias 1500010O08Rik; AI451582; AI503389; AI504772; BOS_19123; BOS_19126; CDCBM4; centrosome marker; gamma tubulin; Gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; Gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; XELAEV_18002398mg; xgam; zgc:55894
Gene Symbols TUBG1, Tubg2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Tubulin gamma-1 chain.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27175, 7283
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.