Learn More
gamma tubulin Mouse anti-Canine, Human, Mouse, Rat, Clone: 3F9H8, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663201IG150UL

Description
This antibody can label the centrosome structure.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.Specifications
gamma tubulin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23258, P83887, P83888, Q9GKK5 | |
TUBG1 | |
gamma tubulin Fusion Protein Ag23913 | |
150 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3F9H8 | |
Unconjugated | |
TUBG1 | |
Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma | |
Mouse | |
Protein A | |
RUO | |
103733, 252921, 403695, 7283 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.