Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gamma-Tubulin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563067
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3067 Supplier Bethyl Laboratories, Inc Supplier No. A302630A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, Bovine and Dog

The gamma-tubulin (TUBG1; relative molecular weight about 48 kDa) is a minor member of tubulin family (less that 0.01% of tubulin dimer). The gamma-tubulin ring structures, however, serve to provide structural primer for initiation of microtubular nucleation and growth, thereby being crutial for microtubule-based cellular processes, above all for mitotic spindle formation. In animal cells, a center of microtubule organization is the centrosome composed of a pair of cylindrical centrioles surrounded by fibrous pericentriolar material containing gamma-tubulin. Formation of the mitotic spindle is preceded by duplication of centrosome during S phase. Before mitosis, both centrosomes increase their microtubule nucleation capacity and form two microtuble asters that are pushed apart from each other by the forces of motor proteins associated at the microtubule surface.

Specifications

Antigen Gamma-Tubulin
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TUBG1
Gene Accession No. P23258
Gene Alias centrosome marker
Gene Symbols TUBG1
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7283
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.