Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109754 Supplier Invitrogen™ Supplier No. PA5109754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSAPSATPIF APGENCSPAW GAAPAAYDAA DTHLRILGKP VMERWETPYM HALAAAASSK GGRVLEVGFG MAIAASKVQE APIDEHWIIE CNDGVFQRLR DWAPRQTHKV IPLKGLWEDV APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKNHAFRLL KPGGVLTYCN LTSWGELMKS KYSDITIMFE ETQVPALLEA GFRRENIRTE VMALVPPADC RYYAFPQMIT PLVTKG.

GAMT is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in its gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals.The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAMT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GAMT
Gene Accession No. O35969, P10868, Q14353
Gene Alias AA571402; CCDS2; epididymis secretory protein Li 20; GAMT; GMT; guanidinoacetate methyltransferase; guanidinoacetate N-methyltransferase; HEL-S-20; hypothetical protein LOC515270; PIG2; Spintz1; TP53I2; zgc:123136; zgc:123136 protein
Gene Symbols GAMT
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14431, 25257, 2593
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.