Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561982 Supplier Invitrogen™ Supplier No. PA561982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACGVAMELYV FGGVRSREDA QGSEMVTCKS EFYHDEFKRW IYLNDQNLCI PASSSFVYGA VPIGASIYVI GDLDTGTNYD YVREFKRSTG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAN
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAN
Gene Accession No. Q9H2C0
Gene Alias A330045G18; gan; GAN1; giant axonal neuropathy; giant axonal neuropathy (gigaxonin); gigaxonin; kelch-like family member 16; kelch-like protein 16; KLHL16
Gene Symbols GAN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAN. Recombinant protein control fragment (Product #RP-98557).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.