Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570399
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
Specifications
GAN | |
Polyclonal | |
Unconjugated | |
GAN | |
A330045G18; gan; GAN1; giant axonal neuropathy; giant axonal neuropathy (gigaxonin); gigaxonin; kelch-like family member 16; kelch-like protein 16; KLHL16 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8139 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2C0 | |
GAN | |
synthetic peptide directed towards the middle region of human GAN. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction