Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 143051AP150UL

Description
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.Specifications
GAN | |
Polyclonal | |
Unconjugated | |
GAN | |
GAN, GAN1, gigaxonin, Kelch like protein 16, KLHL16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209239, 307893, 8139 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CA72, Q9H2C0 | |
GAN | |
GAN Fusion Protein Ag5421 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction