Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GANAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582943 Supplier Invitrogen™ Supplier No. PA582943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYQPFFRAHA HLDTGRREPW LLPSQHNDII RDALGQRYSL LPFWYTLLYQ AHREGIPVMR PLWVQYPQDV TTFNIDDQYL LGDALLVHPV SDSGAHGVQV YLPGQGEVWY DIQSYQKHHG PQTLYLPVTL SSIPVFQ Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 87%.

This gene encodes the alpha subunit of glucosidase II and a member of the glycosyl hydrolase 31 family of proteins. The heterodimeric enzyme glucosidase II plays a role in protein folding and quality control by cleaving glucose residues from immature glycoproteins in the endoplasmic reticulum. Expression of the encoded protein is elevated in lung tumor tissue and in response to UV irradiation. Mutations in this gene cause autosomal-dominant polycystic kidney and liver disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GANAB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ganab
Gene Accession No. Q14697
Gene Alias alpha glucosidase 2 alpha neutral subunit; alpha-glucosidase 2; AU042638; carbohydrate processing enzyme of the endoplasmic reticulum; G2an; GANAB; GIIA; glucosidase II alpha subunit; glucosidase II alpha-subunit; glucosidase II subunit alpha; glucosidase, alpha; neutral AB; GLUII; KIAA0088; mKIAA0088; neutral AB; neutral alpha-glucosidase AB
Gene Symbols Ganab
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GANAB. Recombinant protein control fragment (Product #RP-90011).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.