Learn More
Invitrogen™ GANAB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589484
Description
Immunogen sequence: MTRFRIDELE PRRPRYRVPD VLVADPPIAR LSVSGRDENS VELTMAEGPY KIILTARPFR LDLLEDRSLL LSVNARGLLE FEHQRAPRVS QGSKDPAEGD GAQPEETPRD GDKPEETQGK AEKDEPGAWE ETFKTHSDSK PYGPMSVGLD FSLPGMEHVY GIPEHADNLR LKVTEGGEPY RLYNLDVFQY ELYNPMALYG SVPVLLAHNP HRDLGIFWLN AAETWVDISS NTAGKTLFGK MMDYLQGSGE TPQTDVRWMS; Positive Samples: U-87MG, Raji, HepG2, HeLa, Mouse brain, Mouse liver, Rat brain; Cellular Location: Endoplasmic reticulum, Golgi apparatus, Melanosome.
This gene encodes the alpha subunit of glucosidase II and a member of the glycosyl hydrolase 31 family of proteins. The heterodimeric enzyme glucosidase II plays a role in protein folding and quality control by cleaving glucose residues from immature glycoproteins in the endoplasmic reticulum. Expression of the encoded protein is elevated in lung tumor tissue and in response to UV irradiation. Mutations in this gene cause autosomal-dominant polycystic kidney and liver disease.
Specifications
GANAB | |
Polyclonal | |
Unconjugated | |
Ganab | |
alpha glucosidase 2 alpha neutral subunit; alpha-glucosidase 2; AU042638; carbohydrate processing enzyme of the endoplasmic reticulum; G2an; GANAB; GIIA; glucosidase II alpha subunit; glucosidase II alpha-subunit; glucosidase II subunit alpha; glucosidase, alpha; neutral AB; GLUII; KIAA0088; mKIAA0088; neutral AB; neutral alpha-glucosidase AB | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14376, 23193, 293721 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q14697, Q8BHN3 | |
Ganab | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human GANAB (NP_0012651231). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.