Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ganglioside GD2 Monoclonal Antibody (14G2a), Abbomax

Catalog No. 502204281
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4281 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4281 Supplier Abbomax Inc Supplier No. 604700
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Positive control: Nerve cells Cellular location: Membrane.

Ganglioside GD2 Monoclonal antibody specifically detects Ganglioside GD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Ganglioside GD2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14G2a
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene B4GALNT1
Gene Accession No. Q00973
Gene Alias (N-acetylneuraminyl)-galactosylglucosylceramide; 4933429D13Rik; B4galnt1; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta1,4GalNAC-T; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta-1,4-N-acetyl-galactosaminyltransferase 1; beta-4N-acetylgalactosaminyltransferase; GALGT; Galgt1; GalNAcT; Gal-NAc-T; GalNAc-T; GD2 synthase, GM2 synthase; Ggm2; Ggm-2; GM2/GD2 synthase; SIAT2; SPG26; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T); UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide-beta-1, 4-N-acetylgalactosaminyltransferase
Host Species Mouse
Immunogen Enriched cell membrane of Neuroblastoma cell lysate
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.