Learn More
Invitrogen™ Ganglioside GM2 Chimeric Recombinant Rabbit Monoclonal Antibody (L55-81)

Description
Specificity: L55-81 binds specifically to GM2 ganglioside; this antibody shows no reactivity towards GM1, GM3, GD2 or normal tissues- the L55-81 epitope contains both a GalNAc and sialic acid bound to lactose. Ganglioside GM2 (GalNAcBeta1-4(NeuNAcalpha2-3)GalBeta1-4GlcBeta-1ceramide) is a glycosphingolipid present on the cell surface and a major tumor-differentiation antigen in human melanoma and hepatoma.
Specifications
Specifications
| Antigen | Ganglioside GM2 Chimeric |
| Applications | ELISA, Functional Assay, Immunohistochemistry, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | L55-81 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | ALB |
| Gene Accession No. | P02769 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.