Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gankyrin Antibody (OTI3F6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271267MFV450
Description
Gankyrin Monoclonal antibody specifically detects Gankyrin in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Gankyrin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI3F6 | |
Flow Cytometry | |
26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
Full length human recombinant protein of human PSMD10 (NP_002805) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
5716 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction