Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gankyrin Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001834PCP
Description
Gankyrin Polyclonal antibody specifically detects Gankyrin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)Specifications
Gankyrin | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular) | |
26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
A synthetic peptide made to an internal portion of the human Gankyrin protein sequence (between residues 100-200). [UniProt# O75832] | |
0.1 mL | |
Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
5716 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Flow Cytometry (Intracellular Staining) | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction