Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GANP Monoclonal Antibody (1H3)
GREENER_CHOICE

Catalog No. PIMA520547
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520547 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520547 Supplier Invitrogen™ Supplier No. MA520547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: VRVARCCEDV CAHLVDLFLV EEIFQTAKET LQELQCFCKY LQRWREAVTA RKKLRRQMRA FPAAPCCVDV SDRLRALAPS AECPIAEENL ARGLLDLGHA.

POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GANP
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MCM3AP
Gene Accession No. O60318
Gene Alias 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; GC-associated DNA primase; germinal center-associated nuclear protein; germinal-center associated nuclear protein; germinal-centre associated nuclear protein; KIAA0572; MAP80; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP; minichromosome maintenance complex component 3 associated protein; minichromosome maintenance deficient 3 (S. cerevisiae) associated protein; minichromosome maintenance deficient 3 associated protein; minichromosome maintenance deficient 3-associated protein; mKIAA0572; nuclear protein GANP; SAC3
Gene Symbols MCM3AP
Host Species Mouse
Immunogen MCM3AP (AAH13285, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.