Learn More
Invitrogen™ GANP Polyclonal Antibody

Description
The antibody detects endogenous levels of total GANP / MCM3AP protein.
Specifications
Specifications
| Antigen | GANP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MCM3AP |
| Gene Accession No. | O60318, Q9WUU9 |
| Gene Alias | 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; GC-associated DNA primase; germinal center-associated nuclear protein; germinal-center associated nuclear protein; germinal-centre associated nuclear protein; KIAA0572; MAP80; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP; minichromosome maintenance complex component 3 associated protein; minichromosome maintenance deficient 3 (S. cerevisiae) associated protein; minichromosome maintenance deficient 3 associated protein; minichromosome maintenance deficient 3-associated protein; mKIAA0572; nuclear protein GANP; SAC3 |
| Gene Symbols | MCM3AP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.