Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GANP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583220 Supplier Invitrogen™ Supplier No. PA583220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDWYDFVWNR TRGIRKDITQ QHLCDPLTVS LIEKCTRFHI HCAHFMCEEP MSSFDAKINN ENMTKCLQSL KEMYQDLRNK GVFCASE Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GANP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCM3AP
Gene Accession No. O60318
Gene Alias 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; GC-associated DNA primase; germinal center-associated nuclear protein; germinal-center associated nuclear protein; germinal-centre associated nuclear protein; KIAA0572; MAP80; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP; minichromosome maintenance complex component 3 associated protein; minichromosome maintenance deficient 3 (S. cerevisiae) associated protein; minichromosome maintenance deficient 3 associated protein; minichromosome maintenance deficient 3-associated protein; mKIAA0572; nuclear protein GANP; SAC3
Gene Symbols MCM3AP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCM3AP. Recombinant protein control fragment (Product #RP-105881).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.