Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GANP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564334
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4334 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4334 Supplier Bethyl Laboratories, Inc Supplier No. A303128AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GANP (germinal center-associated nuclear protein) is up-regulated selectively in germinal centers and carries an MCM-binding domain in the carboxyl-terminal side. It was found that a region (from 414 to 550 aa) in GANP molecule that is slightly similar to the known DNA-primase component p49. The recombit GANP fragment covering this region synthesizes RNA primers for extension by DNA polymerase I with single-stranded DNA templates in vitro. GANP DNA-primase activity is controlled by phosphorylation at Ser(502) that is induced by CD40-mediated signaling in vitro and in the germinal center B cells stimulated with antigen in vivo. Overexpression of ganp cDNA in Daudi B cells caused the increased DNA synthesis more than the levels of the mock-transfectants. These evidences suggested that the novel DNA-primase GANP is involved in regulation of cell proliferation of antigen-driven B cells in germinal centers.

Specifications

Antigen GANP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MCM3AP
Gene Accession No. O60318
Gene Alias GANP, MAP80, 80 kDa MCM3-associated protein, MCM3 import protein, MCM3 minichromosome maintece deficient 3 associated protein, germinal center-associated nuclear protein, minichromosome maintece deficient 3-associated protein
Gene Symbols MCM3AP
Host Species Rabbit
Immunogen Between 1930 and 1980
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8888
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.