Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GANP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 110541AP150UL

Description
POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.Specifications
GANP | |
Polyclonal | |
Unconjugated | |
MCM3AP | |
80 kDa MCM3 associated protein, GANP, KIAA0572, MAP80, MCM3AP, Protein GANP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294339, 54387, 8888 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60318, Q9WUU9 | |
Mcm3ap | |
GANP Fusion Protein Ag1486 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction