Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAP-43 Antibody (1E3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192713R
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
GAP-43 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1000 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein. | |
0.1 mL | |
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
2596 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction