Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAP-43 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25466825UL
Description
GAP-43 Polyclonal antibody specifically detects GAP-43 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GAP-43 | |
Polyclonal | |
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°CC long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
GAP43 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:KEEPKQADVPAAVTAAAATTPAAEDAAAKATAQPPTETGESSQAEENIEAVDETKPKESARQDE | |
25 μL | |
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience | |
2596 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction