Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP-43 Antibody, Novus Biologicals™
SDP

Catalog No. p-5065541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300143SS 0.025 mL
NB300143 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300143SS Supplier Novus Biologicals Supplier No. NB300143SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 75 publications

GAP-43 Polyclonal specifically detects GAP-43 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Equine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen GAP-43
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, Flow Cytometry, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen 1-2 ug/ml
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Alias Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen C-terminal peptide of rodent GAP43, KEDPEADQEHA coupled to KLH.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2596
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Drosophila, Equine, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.