Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAP43 Recombinant Rabbit Monoclonal Antibody (8E7C8)
GREENER_CHOICE

Catalog No. PIMA535234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535234 Supplier Invitrogen™ Supplier No. MA535234
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ETESATKAST DNSPSSKAED APAKEEPKQA DVPAAVTAAA ATTPAAEDAA AKATAQPPTE TGESSQAEEN IEAVDETKPK ESARQDEGKE EEPEADQEHA.

GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAP43
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 8E7C8
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene GAP43
Gene Accession No. P06837, P07936, P17677
Gene Alias Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14432, 2596, 29423
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.