Learn More
Invitrogen™ GAP43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534943
Description
Recommended positive controls: SK-N-SH, IMR32, SK-N-AS, mouse fetal brain, mouse hippocampus, rat hippocampus, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
GAP43 | |
Polyclonal | |
Unconjugated | |
GAP43 | |
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14432, 2596, 29423 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P06837, P07936, P17677 | |
GAP43 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of rat GAP43. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.