Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAP43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585949 Supplier Invitrogen™ Supplier No. PA585949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAP43 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAP43
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene GAP43
Gene Accession No. P06837, P07936, P17677
Gene Alias Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen Synthetic peptide within mouse GAP43 aa 178-227.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14432, 2596, 29423
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.