Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAP43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143607
Description
GAP43 Polyclonal Antibody for Western Blot, ICC/IF, IHC
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile growth cones that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
GAP43 | |
Polyclonal | |
Unconjugated | |
GAP43 | |
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100061125, 100623423, 14432, 2596, 281777, 29423 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 5mM sodium azide | |
P06836, P06837, P07936, P17677 | |
GAP43 | |
C-terminal peptide of rodent GAP43, KEDPEADQEHA coupled to KLH (aa 216-226). | |
100 μL | |
Primary | |
Human, Mouse, Rat, Bovine, Pig, Horse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction