Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAP43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519470
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519470 Supplier Invitrogen™ Supplier No. PA519470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

For Western Blot, this antibody has non-specific bands at 100 kDa and 50 kDa. Not suitable for use with Immunohistochemistry (Paraffin). This antibody is predicted to react with human and chimpanzee based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.

GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile growth cones that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAP43
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene GAP43
Gene Accession No. P06837, P07936
Gene Alias Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 200 to the C-terminus of Rat GAP43.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14432, 29423
Target Species Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.