Learn More
Invitrogen™ GAP43 Polyclonal Antibody, DyLight™ 550

Description
This antibody does not react with xenopus samples. Suggested positive control: embryonic nervous tissue, brain, injured nerve. Immunogen sequence: KEDPEADQEH A.
Specifications
Specifications
| Antigen | GAP43 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.57 mg/mL |
| Conjugate | DyLight 550 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | GAP43 |
| Gene Accession No. | P06836, P06837, P07936, P17677, P35001 |
| Gene Alias | Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 |
| Gene Symbols | GAP43 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.