Learn More
Invitrogen™ GAP43 Polyclonal Antibody, DyLight™ 650
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA522896
Catalog No. PIPA522896
Only null left
Add to Cart
Description
Description
This antibody does not react with xenopus samples. Suggested positive control: embryonic nervous tissue, brain, injured nerve. Immunogen sequence: KEDPEADQEH A.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAP43
Polyclonal
DyLight 650
GAP43
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Rabbit
Antigen affinity chromatography
RUO
100061125, 100623423, 14432, 2596, 281777, 29423, 427955, 478572
4°C, store in dark
Liquid
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.70 mg/mL
50mM sodium borate with 0.05% sodium azide
P06836, P06837, P07936, P17677, P35001
GAP43
C-terminal peptide of rodent GAP43 coupled to KLH.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Chicken, Drosophila
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.