Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH-2 Antibody (Hs-8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500456
Description
GAPDH-2 Monoclonal specifically detects GAPDH-2 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
GAPDH-2 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, Flow Cytometry 3 to 12 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
GAPDHS | |
This GAPDH-2 antibody was developed against the full length human protein extracted from sperms. | |
0.1 mg | |
Primary | |
The antibody Hs-8 reacts with GAPDHS, the sperm-specific glyceraldehyde phosphate dehydrogenase, previously identified under the general name “intra-acrosomal protein”. | |
Store at 4C. Do not freeze. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Hs-8 | |
Unconjugated | |
EC 1.2.1, EC 1.2.1.12, GAPD2HSD-35, GAPDH2, GAPDH-2glyceraldehyde-3-phosphate dehydrogenase, testis-specific, GAPDS, glyceraldehyde-3-phosphate dehydrogenase, spermatogenic, Spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2, Spermatogenic glyceraldehyde-3-phosphate dehydrogenase | |
Mouse | |
Affinity Purified | |
RUO | |
26330 | |
Human, Pig | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction