Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP303522100UL

 View more versions of this product

Catalog No. NB092569


Only null left
Add to Cart

Description

Description

GAPDH-2 Polyclonal specifically detects GAPDH-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GAPDH-2
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
EC 1.2.1, EC 1.2.1.12, GAPD2HSD-35, GAPDH2, GAPDH-2glyceraldehyde-3-phosphate dehydrogenase, testis-specific, GAPDS, glyceraldehyde-3-phosphate dehydrogenase, spermatogenic, Spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2, Spermatogenic glyceraldehyde-3-phosphate dehydrogenase
Recombinant fusion protein containing a sequence corresponding to amino acids 179-408 of human GAPDH-2 (NP_055179.1).
100 μL
Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
26330
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.