Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH Antibody (13H12), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227103AF405
Description
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Drosophila, Primate, Monkey, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GAPDH | |
| Monoclonal | |
| Alexa Fluor 405 | |
| P04406 | |
| GAPDH | |
| Amino acids between 275 and 325 of glyceraldehyde 3-phosphate dehydrogenase protein were used as the immunogen for this GAPDH antibody. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reacts with Human, Primates/Monkey, Mouse and Drosophila. Based upon 91% sequence similarity with immunogen, this antibody is predicted to react with Guinea Pig, Sheep, Squirrel, Porcine/Pig, Ferret, Canine/Dog/Cat, Bovine, Reptile / Rattlesnake and several other species. Immunogen shows 82% similarity to Xenopus and Zebrafish. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 13H12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| Mouse | |
| 36 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| 2597 | |
| Human, Mouse, Rat, Drosophila, Primate, Monkey, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction