Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH Antibody (GA1R) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$196.10 - $512.00
Specifications
Antigen | GAPDH |
---|---|
Clone | GA1R |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000 - 1:10000, Simple Western, Flow Cytometry 2 μg, ELISA 1:100 - 1:2000, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Bacteria, Chicken, Hamster, Insect, Rabbit, Yeast samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
GAPDH | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
PBS (pH 7.2) with No Preservative | |
2597 | |
This GAPDH protein was developed against recombinant GAPDH | |
Primary | |
This GAPDH antibody detects GAPDH from BL-21 bacteria, Sf9 insect, Saccharomyces cerevisiae (yeast), human, mouse, rat, rabbit, hamster, and chicken samples. This product has been successfully used in Western blot, ELISA, and immunofluorescence. |
GA1R | |
Western Blot 1:1000 - 1:10000, Simple Western, Flow Cytometry 2 μg, ELISA 1:100 - 1:2000, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bacteria, Chicken, Hamster, Insect, Rabbit, Yeast | |
aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
GAPDH | |
IgG1 | |
Protein A purified | |
36 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title