Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 24 publications
$640.48
Specifications
| Antigen | GAPDH |
|---|---|
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:2000-1:10000, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GAPDH | |
| Western Blot 1:2000-1:10000, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| GAPDH | |
| IgG | |
| Affinity Purified | |
| Reacts with Human, Mouse, Rat and Primate sp. Based upon 100% sequence homology, this antibody is expected to react with Hamster (Chinese hamster, Golden Hamster), Canines, Ferret, Squirrel, Pig/Porcine, Bovine, Sheep, Rabbit, Xenopus and other amphibians such as Turtle, Avian species (including Chicken, Turkey, Sparrows, Duck), Zebrafish and several other fishes. |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Chicken, Primate | |
| P04406 | |
| 2597 | |
| This GAPDH antibody was developed against an epitope between residues 150 and 200 of human GAPDH using the numbering given in entry NP_002037.2 (GeneID 2597). | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 36 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title