Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Mouse, anti-Human, CoraLite 594, Clone: 1E6D9, Proteintech
SDP

Catalog No. 502257553 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7553 Supplier Proteintech Group Inc Supplier No. CL59460004100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The GAPDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GAPDH. This antibody recognizes human, mouse, rat, zebrafish, yeast, plant antigen. The GAPDH antibody has been validated for the following applications: FC (Intra), WB analysis. This antibody is coralite 594.

Specifications

Antigen GAPDH
Applications Flow Cytometry (Intracellular Staining), Western Blot
Clone 1E6D9
Concentration 1000 μg/mL
Conjugate CoraLite 594
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3
Host Species Mouse
Immunogen GAPDH fusion protein Ag0766
Purification Method Protein A Purification
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2597
Target Species Human, Mouse, Plant, Rat, Yeast
Content And Storage Store at -20°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.