Learn More
GAPDH Rabbit, Clone: RM114, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of human GAPDH.
Supplier: Abnova Corporation MAB12829
Description
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq]
Specifications
GAPDH | |
Monoclonal | |
Unconjugated | |
ELISA Flow Cytometry Immunocytochemistry (1:200) Immunohistochemistry (1:400-1:4000) Immunoprecipitation (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
G3PD/GAPD/MGC88685 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
2597 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM114 | |
Rabbit monoclonal antibody raised against synthetic peptide of human GAPDH. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
GAPDH | |
A synthetic peptide corresponding to C-terminus region of human GAPDH. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.