Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GAPDH Recombinant Protein Antigen
SDP

Catalog No. NBP248721X Shop All R&D Systems Products
Change view
Click to view available options
Molecular Weight (g/mol):
24 kDa
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Molecular Weight (g/mol)
NBP248721X 24 kDa
NBP259779X None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP248721X Supplier Novus Biologicals™ Supplier No. NBP248721PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GAPDH The GAPDH Recombinant Protein Antigen is derived from E. coli. The GAPDH Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name GAPDH Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Description Blocking peptide for Mfr. No. NBP2-48721
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbol GAPDH
Label Type Unlabeled
Molecular Weight (g/mol) 24 kDa
Product Type Recombinant Protein Antigen
Quantity 0.1 mL
Regulatory Status RUO
Source E.coli
Specific Reactivity Human
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.