Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAPDHS Monoclonal Antibody (Hs-8), FITC
GREENER_CHOICE

Catalog No. PIMA544094
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544094 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544094 Supplier Invitrogen™ Supplier No. MA544094
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody reacts with GAPDHS, the sperm-specific glyceraldehyde phosphate dehydrogenase, which is an intra-acrosomal protein.

Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. Because GAPDH is a protein expressed in large amounts and which is required at all times for important 'house keeping' functions, levels of GAPDH mRNA are often measured and used as standards in studies of mRNA expression. Increasingly, scientists are making use of specific antibodies to GAPDH in comparable studies of levels of protein expression. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer's disease, and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington's disease. Associations with actin and tubulin have also been reported. The protein may also have a role in the regulation of apoptosis, and interestingly migrates from the cytoplasm into the nucleus when cells become apoptotic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPDHS
Applications Flow Cytometry
Classification Monoclonal
Clone Hs-8
Concentration 0.06 mg/mL
Conjugate FITC
Formulation TBS with 0.2% BSA and 15mM sodium azide; pH 8
Gene GAPDHS
Gene Accession No. O14556
Gene Alias epididymis secretory protein Li 278; GAPD2; GAPDH S; GAPDH2; gapdh-2; GAPDHS; GAPDS; Gapd-s; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; HEL-S-278; HSD35; HSD-35; LOW QUALITY PROTEIN: glyceraldehyde-3-phosphate dehydrogenase, testis-specific; spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2; Spermatogenic glyceraldehyde-3-phosphate dehydrogenase
Gene Symbols GAPDHS
Host Species Mouse
Immunogen Freshly ejaculated human sperms.
Purification Method Size-exclusion chromatography
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100517097, 26330
Target Species Human, Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.