Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAPDHS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587928 Supplier Invitrogen™ Supplier No. PA587928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASDHISAGAQ RVVISAPSPD APMFVMGVNE NDYNPGSMNI VSNASCTTNC LAPLAKVIHE RFGIVEGLMT TVHSYTATQK TVDGPSRKAW RDGRGAHQNI IPASTGAAKA VTKVIPELKG KLTGMAFRVP TPDVSVVDLT CRLAQPAPYS AIKEAVKAAA KGPMAGILAY TEDEVVSTDF LGDTHSSIFD AKAGIALNDN FVKLISWYDN EYGYSHRVVD LLRYMFSRDK; Positive Samples: HT-1080, U-251MG, SW620, MCF-7, Mouse liver, Rat testis; Cellular Location: Cytoplasm.

Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. Because GAPDH is a protein expressed in large amounts and which is required at all times for important 'house keeping' functions, levels of GAPDH mRNA are often measured and used as standards in studies of mRNA expression. Increasingly, scientists are making use of specific antibodies to GAPDH in comparable studies of levels of protein expression. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer's disease, and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington's disease. Associations with actin and tubulin have also been reported. The protein may also have a role in the regulation of apoptosis, and interestingly migrates from the cytoplasm into the nucleus when cells become apoptotic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPDHS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GAPDHS
Gene Accession No. O14556, Q64467, Q9ESV6
Gene Alias epididymis secretory protein Li 278; GAPD2; GAPDH S; GAPDH2; gapdh-2; GAPDHS; GAPDS; Gapd-s; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; HEL-S-278; HSD35; HSD-35; LOW QUALITY PROTEIN: glyceraldehyde-3-phosphate dehydrogenase, testis-specific; spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2; Spermatogenic glyceraldehyde-3-phosphate dehydrogenase
Gene Symbols GAPDHS
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14447, 26330, 66020
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.