Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552785 Supplier Invitrogen™ Supplier No. PA552785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATRFTLPRFL QRRSSRRKVC TKTFLGPRII GLRHEISVET QDHKSAVRGN NTHDNYENVE AGPPKAKGKT DKELYENTGQ SNFEEHIYGN ETSSDYYNFQ KPRPSEVPQD ED Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.

Negatively regulates B-cell proliferation following stimulation through the B-cell receptor. May play an important role in maintenance of marginal zone (MZ) B-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAPT
Gene Accession No. Q8N292
Gene Alias C5orf29; GAPT; GRB2-binding adaptor protein, transmembrane; GRB2-binding transmembrane adaptor; growth factor receptor-bound protein 2-binding adapter protein, transmembrane; Protein GAPT
Gene Symbols GAPT
Host Species Rabbit
Immunogen Recombinant protein fragment of Human GAPT (Product #RP-90481)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.