Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAPT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139741
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ATRFTLPRFL QRRSSRRKVC TKTFLGPRII GLRHEISVET QDHKSAVRGN NTHDNYENVE AGPPKAKGKT DKELYENTGQ SNFEEHIYGN ETSSDYYNFQ KPRPSEVPQD ED.
Negatively regulates B-cell proliferation following stimulation through the B-cell receptor. May play an important role in maintenance of marginal zone (MZ) B-cells.
Specifications
GAPT | |
Polyclonal | |
Unconjugated | |
GAPT | |
C5orf29; GAPT; GRB2-binding adaptor protein, transmembrane; GRB2-binding transmembrane adaptor; growth factor receptor-bound protein 2-binding adapter protein, transmembrane; Protein GAPT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
202309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N292 | |
GAPT | |
Recombinant protein corresponding to Human GAPT. Recombinant protein control fragment (Product #RP-90481). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction