Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Garden Guide Kit



Click to view available options
Product Type:
Garden Guide Kit
Reagent Refill
Description
Includes
Reagent Refill
Measures nitrogen, phosphorus, potassium and soil pH.
- New hinged carrying case with foam liner hold all components securely
- Includes the LaMotte Soil Handbook, Garden Guide Manual, instructions, and free downloadable resource, A Study of Soil Science handbook
- Grades 5+
Specifications
Specifications
Product Type | Reagent Refill |
Grade | 5 |
For Use With (Application) | Soil science education or garden analysis by measuring nitrogen, phosphorus, potassium and soil ph |
Class Size | 1 Class |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction