Learn More
Eisco™ Garnet Mineral Specimen
SCI_ED
Catalog No. S27599 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27599 12/Pk.
S27577 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27599 Supplier Eisco™ Supplier No. ESNG0070PK12
Add to Cart
Add to Cart
Garnet forms under intense heat and pressure conditions, making it a common mineral in mountainous terranes.
  • Correlates well to Next Generation Science Standards Evidence while Garnet does come in a variety of colors, this specimen is a deep red color
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Great for lessons in metamorphic facies & silicate minerals.
Specifications
Product Type Mineral
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg