Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARNL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982772
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2772 Supplier Bioss Supplier No. BS13286R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.

Specifications

Antigen GARNL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RALGAPA1
Gene Accession No. Q6GYQ0
Gene Alias 2310003F20Rik; 4930400K19Rik; AI563624; GAP-related interacting partner to E12; GAP-related interacting protein to E12; GAP-related-interacting partner to E12; GARNL1; GRIPE; GTPase activating RANGAP domain-like 1; GTPase activating Rap/RanGAP domain-like 1; GTPase-activating Rap/Ran-GAP domain-like 1; GTPase-activating RapGAP domain-like 1; KIAA0884; mKIAA0884; p240; Ral GTPase activating protein catalytic alpha subunit 1; Ral GTPase activating protein, alpha subunit 1; Ral GTPase activating protein, alpha subunit 1 (catalytic); ral GTPase-activating protein alpha subunit 1; Ral GTPase-activating protein subunit alpha-1; Ralgapa1; RalGAPalpha1; Tuberin-like protein 1; tulip 1 protein; Tulip1
Gene Symbols RALGAPA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GARNL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253959
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.