Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARNL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982771
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2771 Supplier Bioss Supplier No. BS13285R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GARNL3 is a protein coding gene. Gene ontology (GO) annotation include intracellular membrane-bounded organelle.

Specifications

Antigen GARNL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GARNL3
Gene Accession No. Q5VVW2
Gene Alias bA356B19.1; GARNL3; GTPase activating RANGAP domain-like 3; GTPase activating Rap/RanGAP domain like 3; GTPase activating Rap/RanGAP domain-like 3; GTPase-activating Rap/Ran-GAP domain-like protein 3
Gene Symbols GARNL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GARNL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84253
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.